ENA410542455 Enamine

2-chloro-5-(propan-2-yl)pyridine

  • MDL number MFCD20265780
  • Cas Number : 68700-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119097
Availability details for
Quantity
  • Availability
  • Add to Cart