ENA410542488 Enamine

1-methyl-1H-pyrazolo[4,3-c]pyridin-3-amine

  • MDL number MFCD26407538
  • Cas Number : 1556454-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125400
Availability details for
Quantity
  • Availability
  • Add to Cart