ENA410542493 Enamine

5H,6H,7H-cyclopenta[c]pyridazin-3-amine

  • MDL number MFCD16871461
  • Cas Number : 1342288-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127543
Availability details for
Quantity
  • Availability
  • Add to Cart