ENA410542496 Enamine

1-(4H-1,2,4-triazole-3-sulfonyl)piperazine

  • MDL number MFCD26407542
  • Cas Number : 1797821-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128175
Availability details for
Quantity
  • Availability
  • Add to Cart