ENA410542511 Enamine

3-benzyl-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-6-amine

  • MDL number MFCD26407549
  • Cas Number : 1708398-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129792

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129792
Availability details for
Quantity
  • Availability
  • Add to Cart