ENA410542513 Enamine

5-bromopent-2-yne

  • MDL number MFCD20621213
  • Cas Number : 18719-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129872

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-129872
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart