ENA410542525 Enamine

2-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-isoindole-5-carboxylic acid

  • MDL number MFCD08234862
  • Cas Number : 149353-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130440
Availability details for
Quantity
  • Availability
  • Add to Cart