ENA410542530 Enamine

5-(difluoromethoxy)pyridine-2-carbonitrile

  • MDL number MFCD14698404
  • Cas Number : 1206979-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38...40
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131314
Availability details for
Quantity
  • Availability
  • Add to Cart