ENA410542533 Enamine

tert-butyl N-{3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]propyl}carbamate

  • MDL number MFCD26407568
  • Cas Number : 1797919-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131484

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131484
Availability details for
Quantity
  • Availability
  • Add to Cart