ENA410542544 Enamine

2-[(4-nitrophenyl)methyl]butanoic acid

  • MDL number MFCD12794555
  • Cas Number : 1248983-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-45
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132403
Availability details for
Quantity
  • Availability
  • Add to Cart