ENA410542545 Enamine

2-(3,5-dimethoxy-2-nitrophenyl)acetonitrile

  • MDL number MFCD26407588
  • Cas Number : 93427-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132501

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132501
Availability details for
Quantity
  • Availability
  • Add to Cart