ENA410542549 Enamine

2-chloro-N-(3,5-dichlorophenyl)-2-phenylacetamide

  • MDL number MFCD03964201
  • Cas Number : 726157-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01871
Availability details for
Quantity
  • Availability
  • Add to Cart