ENA410542554 Enamine

5-ethoxy-8-nitroquinoline

  • MDL number MFCD26407600
  • Cas Number : 1803609-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133271
Availability details for
Quantity
  • Availability
  • Add to Cart