ENA410542556 Enamine

3,6-dihydro-2H-pyran-4-carboxylic acid

  • MDL number MFCD20040387
  • Cas Number : 99338-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133424
Availability details for
Quantity
  • Availability
  • Add to Cart