ENA410542565 Enamine

1-(3-methyl-1-benzothiophen-2-yl)ethan-1-ol

  • MDL number MFCD12047709
  • Cas Number : 69994-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133795
Availability details for
Quantity
  • Availability
  • Add to Cart