ENA410542570 Enamine

2,6-dichloro-7H-purin-8-ol

  • MDL number MFCD07783218
  • Cas Number : 98027-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134125
Availability details for
Quantity
  • Availability
  • Add to Cart