ENA410542577 Enamine

5-(2-methylphenyl)-1,2,4-thiadiazol-3-amine

  • MDL number MFCD11655558
  • Cas Number : 1086385-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134505
Availability details for
Quantity
  • Availability
  • Add to Cart