ENA410542578 Enamine

5-(2-chlorophenyl)-1,2,4-thiadiazol-3-amine

  • MDL number MFCD11655563
  • Cas Number : 42053-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134528
Availability details for
Quantity
  • Availability
  • Add to Cart