ENA410542581 Enamine

4-bromo-5-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione

  • MDL number MFCD26407625
  • Cas Number : 732295-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256...258
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134667
Availability details for
Quantity
  • Availability
  • Add to Cart