ENA410542585 Enamine

3-chloro-1aH,6H,6aH-indeno[1,2-b]oxirene

  • MDL number MFCD09026594
  • Cas Number : 116540-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135013
Availability details for
Quantity
  • Availability
  • Add to Cart