ENA410542597 Enamine

4-chloro-3-methoxypyridine-2-carboxylic acid

  • MDL number MFCD09909844
  • Cas Number : 170621-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135352

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135352
Availability details for
Quantity
  • Availability
  • Add to Cart