ENA410542608 Enamine

1,2,3,4-Tetrahydro-acridin-9-ylamine

  • MDL number MFCD00046923
  • Cas Number : 321-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280...284
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02105

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02105
Availability details for
Quantity
  • Availability
  • Add to Cart