ENA410542610 Enamine

(4R)-7-fluoro-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD15529792
  • Cas Number : 1270296-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135824
Availability details for
Quantity
  • Availability
  • Add to Cart