ENA410542616 Enamine

2-(4-methylbenzoyl)-1-benzofuran-3-amine

  • MDL number MFCD01578621
  • Cas Number : 333435-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135927
Availability details for
Quantity
  • Availability
  • Add to Cart