ENA410542618 Enamine

methyl 2-amino-3-hydroxybutanoate hydrochloride

  • MDL number MFCD00070400
  • Cas Number : 2170123-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136012
Availability details for
Quantity
  • Availability
  • Add to Cart