ENA410542622 Enamine

3-(3,4-dichlorobenzoyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD11607100
  • Cas Number : 893383-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136307

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136307
Availability details for
Quantity
  • Availability
  • Add to Cart