ENA410542624 Enamine

ethyl 3-(2,6-dichlorophenyl)-2-methylprop-2-enoate

  • Cas Number : 119424-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136341
Availability details for
Quantity
  • Availability
  • Add to Cart