ENA410542641 Enamine

1-(5-methylpyridin-3-yl)ethan-1-one

  • MDL number MFCD11553496
  • Cas Number : 42972-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136658
Availability details for
Quantity
  • Availability
  • Add to Cart