ENA410542643 Enamine

5-Nitroso-quinolin-8-ol

  • MDL number MFCD00041862
  • Cas Number : 3565-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03092

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-03092
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart