ENA410542647 Enamine

5-bromo-8-chloro-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD21706112
  • Cas Number : 1445948-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136794

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-136794
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart