ENA410542666 Enamine

3-methyl-1-[(pyridin-3-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD26407789
  • Cas Number : 1803571-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137017
Availability details for
Quantity
  • Availability
  • Add to Cart