ENA410542678 Enamine

2-chloro-N-ethyl-N-(2-methylprop-2-en-1-yl)acetamide

  • MDL number MFCD03964595
  • Cas Number : 726153-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04557
Availability details for
Quantity
  • Availability
  • Add to Cart