ENA410542682 Enamine

(3-fluoro-4-methylphenyl)methanol

  • MDL number MFCD01631533
  • Cas Number : 192702-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34...36
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137201

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-137201
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart