ENA410542684 Enamine

methyl 2-cyclopropanecarbonyl-3-(dimethylamino)prop-2-enoate

  • MDL number MFCD18632625
  • Cas Number : 340774-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137210
Availability details for
Quantity
  • Availability
  • Add to Cart