ENA410542687 Enamine

6-methyl-6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-amine

  • MDL number MFCD16090980
  • Cas Number : 1247088-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137287
Availability details for
Quantity
  • Availability
  • Add to Cart