ENA410542693 Enamine

6'-methoxy-3',4'-dihydro-2'H-spiro[imidazolidine-4,1'-naphthalene]-2,5-dione

  • MDL number MFCD03964681
  • Cas Number : 30265-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-04926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04926
Availability details for
Quantity
  • Availability
  • Add to Cart