ENA410542704 Enamine

4-(3-bromopropyl)-1,2-dichlorobenzene

  • MDL number MFCD12024968
  • Cas Number : 29648-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137367
Availability details for
Quantity
  • Availability
  • Add to Cart