ENA410542708 Enamine

4-fluoro-4-phenylpiperidine hydrochloride

  • MDL number MFCD13176614
  • Cas Number : 1056382-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137379

Safety Information

signalword Warning
pcodes P264,P280,P302+P352,P305+P351+P338,P332+P313,P337+P313,P362
MSDS https://www.enaminestore.com/catalog/EN300-137379
pictograms GHS07
hstatements H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart