ENA410542722 Enamine

methyl 2-(3-methylpiperidin-1-yl)-2-oxoacetate

  • MDL number MFCD25978443
  • Cas Number : 1565404-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137488
Availability details for
Quantity
  • Availability
  • Add to Cart