ENA410542727 Enamine

1-benzyl-4-methylpyrrolidin-3-amine

  • MDL number MFCD11858564
  • Cas Number : 74880-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137521
Availability details for
Quantity
  • Availability
  • Add to Cart