ENA410542738 Enamine

N-(2-hydroxypropyl)-N-methylmethanesulfonamide

  • MDL number MFCD16711113
  • Cas Number : 106894-77-3
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-137545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137545
Availability details for
Quantity
  • Availability
  • Add to Cart