ENA410542739 Enamine

2-(2-fluorophenyl)-2-methylpropane-1,3-diol

  • MDL number MFCD21927420
  • Cas Number : 1504111-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137549
Availability details for
Quantity
  • Availability
  • Add to Cart