ENA410542743 Enamine

2-(4-methyl-5-phenyl-4H-1,2,4-triazol-3-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD26407861
  • Cas Number : 1803598-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137554

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137554
Availability details for
Quantity
  • Availability
  • Add to Cart