ENA410542745 Enamine

methyl 2-amino-4,4,4-trifluorobutanoate hydrochloride

  • MDL number MFCD08532483
  • Cas Number : 1007583-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137557
Availability details for
Quantity
  • Availability
  • Add to Cart