ENA410542748 Enamine

tert-butyl N-[1-(2,2,2-trifluoroethyl)piperidin-4-yl]carbamate

  • MDL number MFCD22419909
  • Cas Number : 335653-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137583

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-137583
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart