ENA410542756 Enamine

3-bromo-1-(4-bromo-3-methylphenyl)pyrrolidin-2-one

  • MDL number MFCD18886569
  • Cas Number : 1343739-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137625

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137625
Availability details for
Quantity
  • Availability
  • Add to Cart