ENA410542758 Enamine

2,2-dimethoxy-1-(pyridin-3-yl)ethan-1-amine

  • MDL number MFCD21937688
  • Cas Number : 1521100-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137627
Availability details for
Quantity
  • Availability
  • Add to Cart