ENA410542763 Enamine

(5-cyanofuran-2-yl)methyl acetate

  • MDL number MFCD15144061
  • Cas Number : 89149-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137654

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137654
Availability details for
Quantity
  • Availability
  • Add to Cart