ENA410542768 Enamine

1-methyl-1H-pyrazole-5-sulfonamide

  • MDL number MFCD18426034
  • Cas Number : 918335-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137690

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137690
Availability details for
Quantity
  • Availability
  • Add to Cart