ENA410542769 Enamine

3-(2-methoxyethyl)pyrrolidine

  • MDL number MFCD13562882
  • Cas Number : 1220035-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137691
Availability details for
Quantity
  • Availability
  • Add to Cart